Assay Details
| Gene Symbol: Car9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139305.2 | NP_647466.2 | 4 - 5 | 717 | 71 | |
| GenBank mRNA | AB086322.1 | - | 4 - 5 | 719 | 71 | |
| AJ245857.1 | - | 4 - 5 | 719 | 71 | ||
| AK136579.1 | - | 4 - 5 | 718 | 71 | ||
| AK137719.1 | - | 4 - 5 | 719 | 71 | ||
| AK166403.1 | - | 1 - 2 | 146 | 71 | ||
| BC120544.1 | - | 4 - 5 | 710 | 40129467 | 71 | |
| BC120546.1 | - | 4 - 5 | 710 | 40129469 | 71 | |
| EF122497.1 | - | 4 - 5 | 719 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car9
Genomic Map