Assay Details
| Gene Symbol: Car9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139305.2 | NP_647466.2 | 9 - 10 | 1199 | 67 | |
| GenBank mRNA | AJ245857.1 | - | 9 - 10 | 1201 | 67 | |
| AK136579.1 | - | 9 - 10 | 1200 | 67 | ||
| AK137719.1 | - | 9 - 10 | 1201 | 67 | ||
| AK166403.1 | - | 6 - 7 | 628 | 67 | ||
| BC120544.1 | - | 9 - 10 | 1192 | 40129467 | 67 | |
| BC120546.1 | - | 9 - 10 | 1192 | 40129469 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car9
Genomic Map