Assay Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144491.2 | NP_652762.2 | 8 - 9 | 929 | 82 | |
| GenBank mRNA | AK082812.1 | - | 8 - 9 | 934 | 82 | |
| AK128996.1 | - | 920 | 82 | |||
| AY078170.1 | - | 8 - 9 | 920 | 82 | ||
| BC022738.1 | - | 3 - 4 | 348 | 82 | ||
| BC044663.1 | - | 8 - 9 | 900 | 82 | ||
| BC080761.1 | - | 8 - 9 | 905 | 82 | ||
More Information
Set Membership: |
|

Dph1
Genomic Map