Assay Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144491.2 | NP_652762.2 | 9 - 10 | 1029 | 67 | |
| GenBank mRNA | AK128996.1 | - | 1020 | 67 | ||
| AY078170.1 | - | 9 - 10 | 1020 | 67 | ||
| BC022738.1 | - | 4 - 5 | 448 | 67 | ||
| BC044663.1 | - | 9 - 10 | 1000 | 67 | ||
| BC080761.1 | - | 9 - 10 | 1005 | 67 | ||
More Information
Set Membership: |
|

Dph1
Genomic Map