Assay Details
| Gene Symbol: Ces3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159415.1 | NP_001152887.1 | 1 - 2 | 118 | 85 | |
| NM_144511.2 | NP_653094.2 | 1 - 2 | 118 | 85 | ||
| GenBank mRNA | BC010812.1 | - | 1 - 2 | 108 | 85 | |
| BC019147.1 | - | 1 - 2 | 114 | 5052937 | 85 | |
| BC019926.1 | - | 1 - 2 | 108 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109802 | CB947075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109802 | BC010812.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109802 | BC019147.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109802 | BC019926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109802 | NM_001159415.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830865 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732628 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146117 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109802 | CB947075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109802 | BC010812.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109802 | BC019147.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109802 | BC019926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109802 | NM_001159415.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26131605 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06625582 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146117 | - | - |
| Whole Mouse Genome G4122A | A_51_P393193 | BC010812.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393193 | BC019147.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393193 | BC019926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393193 | NM_001159415.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393193 | NM_144511.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109802 | CB947075.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109802 | BC010812.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109802 | BC019147.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109802 | BC019926.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109802 | NM_001159415.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06757207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26131650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07408645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06852541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07331692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06784764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07203731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06862885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07408638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07001315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06852540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06852534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07408644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06852539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26131577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10574610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574613 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574614 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574608 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574611 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574612 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574607 | - | - |
| Mouse Genome 430 2.0 Array | 1451600_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1451600_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800009416.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009416.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009416.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009416.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009416.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009420.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009420.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009420.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009420.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009420.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009412.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009412.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009412.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009412.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009412.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009414.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009414.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009414.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009414.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009414.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009418.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009418.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009418.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009404.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009404.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009404.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009404.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009404.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009415.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009415.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009415.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009415.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009415.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009411.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009411.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009411.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009411.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009411.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009419.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009419.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009419.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009419.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009419.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009421.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009421.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009421.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009421.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009421.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009405.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009405.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009405.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009405.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009405.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009410.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009410.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009410.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009410.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009410.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009406.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009406.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009406.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009406.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009406.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009408.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009408.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009408.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009408.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009408.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009409.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009409.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009409.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009409.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009409.mm.1 | TC0800001201.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009417.mm.1 | NM_001159415.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009417.mm.1 | NM_001159415.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009417.mm.1 | NM_144511.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009417.mm.1 | NM_144511.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009417.mm.1 | TC0800001201.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451600_PM_s_at | - | - |
| Mouse Expression Set 430 | 1451600_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451600_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451600_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 575060 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6370064 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6760528 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006760528 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254721 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733745 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2733745 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254721 | - | - |
| mouse-6 v1.1 (Illumina) | scl013909.6_35-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0013909.2_193-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6760528 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013909.6_35-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013909.2_193-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370064 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760528 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254721 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2917053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2733745 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2917053 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254721 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733745 | - | - |

Ces3b
Genomic Map