Assay Details
| Gene Symbol: Cpa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144537.3 | NP_653120.1 | 2 - 3 | 653 | 102 | |
| GenBank mRNA | AF466283.1 | - | 2 - 3 | 654 | 102 | |
| AK015256.1 | - | 2 - 3 | 654 | 102 | ||
| BC138114.1 | - | 1 - 2 | 171 | 8861133 | 102 | |
| BC145151.1 | - | 1 - 2 | 171 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpa5
Genomic Map