Assay Details
| Gene Symbol: Cpa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144537.3 | NP_653120.1 | 9 - 10 | 1367 | 81 | |
| GenBank mRNA | AF466283.1 | - | 9 - 10 | 1368 | 81 | |
| AK015256.1 | - | 9 - 10 | 1370 | 81 | ||
| BC138114.1 | - | 8 - 9 | 885 | 8861133 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpa5
Genomic Map