Assay Details
| Gene Symbol: Dlgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144553.2 | NP_653136.2 | 10 - 11 | 1551 | 106 | |
| GenBank mRNA | AB076696.1 | - | 10 - 11 | 1551 | 106 | |
| AK036384.1 | - | 10 - 11 | 1353 | 106 | ||
| AK048470.1 | - | 10 - 11 | 1321 | 106 | ||
| AK085882.1 | - | 10 - 11 | 1332 | 106 | ||
| AK129032.1 | - | 10 - 11 | 1297 | 106 | ||
| AK135820.1 | - | 10 - 11 | 1358 | 106 | ||
| AK147955.1 | - | 10 - 11 | 1321 | 106 | ||
| BC027273.1 | - | 10 - 11 | 1272 | 106 | ||
| BC043924.1 | - | 2 - 3 | 177 | 106 | ||
| BC058087.1 | - | 10 - 11 | 1285 | 6816660 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap5
Genomic Map