Assay Details
| Gene Symbol: Galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144908.3 | NP_659157.1 | 1 - 2 | 144 | 94 | |
| GenBank mRNA | AK088014.1 | - | 1 - 2 | 133 | 94 | |
| BC011428.1 | - | 1 - 2 | 120 | 2631696 | 94 | |
| BC021504.1 | - | 1 - 2 | 145 | 5346678 | 94 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK088014.1,BC011428.1,BC021504.1,NM_144908.3 transcripts.
Set Membership: |
|

Galnt11
Genomic Map