Assay Details
| Gene Symbol: Galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144908.3 | NP_659157.1 | 7 - 8 | 1260 | 96 | |
| GenBank mRNA | AF495528.1 | - | 6 - 7 | 1132 | 96 | |
| AK088014.1 | - | 7 - 8 | 1249 | 96 | ||
| AK161524.1 | - | 7 - 8 | 1170 | 96 | ||
| BC011428.1 | - | 7 - 8 | 1236 | 2631696 | 96 | |
| BC021504.1 | - | 7 - 8 | 1261 | 5346678 | 96 | |
| BC031150.1 | - | 6 - 7 | 1158 | 96 | ||
| BC034184.1 | - | 6 - 7 | 1160 | 4974216 | 96 | |
| BC034185.1 | - | 6 - 7 | 1212 | 4974695 | 96 | |
| BC036143.1 | - | 6 - 7 | 1161 | 4974645 | 96 | |
| BC036145.1 | - | 6 - 7 | 1154 | 4975474 | 96 | |
| Y12435.1 | - | 6 - 7 | 1083 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Galnt11
Genomic Map