Assay Details
| Gene Symbol: Med17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144933.1 | NP_659182.1 | 2 - 3 | 480 | 62 | |
| GenBank mRNA | AK033151.1 | - | 2 - 3 | 486 | 62 | |
| AK038451.1 | - | 2 - 3 | 520 | 62 | ||
| AK049112.1 | - | 2 - 3 | 520 | 62 | ||
| AK142033.1 | - | 2 - 3 | 519 | 62 | ||
| AK151377.1 | - | 2 - 3 | 520 | 62 | ||
| BC004062.1 | - | 1 - 2 | 157 | 62 | ||
| BC020185.1 | - | 2 - 3 | 480 | 4505959 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med17
Genomic Map