Assay Details
| Gene Symbol: Lpcat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145130.2 | NP_660112.1 | 5 - 6 | 600 | 77 | |
| GenBank mRNA | AB294194.1 | - | 5 - 6 | 495 | 77 | |
| AK083687.1 | - | 3 - 4 | 156 | 77 | ||
| AK098090.1 | - | 5 - 6 | 571 | 77 | ||
| AK154062.1 | - | 5 - 6 | 595 | 77 | ||
| AK169452.1 | - | 4 - 5 | 717 | 77 | ||
| AK190594.1 | - | 2 - 3 | 94 | 77 | ||
| AK197070.1 | - | 2 - 3 | 94 | 77 | ||
| AK203927.1 | - | 2 - 3 | 94 | 77 | ||
| AK204330.1 | - | 2 - 3 | 94 | 77 | ||
| AK206831.1 | - | 2 - 3 | 94 | 77 | ||
| AY028317.1 | - | 5 - 6 | 576 | 77 | ||
| BC006753.1 | - | 5 - 6 | 528 | 3709076 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat3
Genomic Map