Assay Details
| Gene Symbol: Aif1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145144.1 | NP_660126.1 | 3 - 4 | 203 | 69 | |
| GenBank mRNA | AB035322.1 | - | 3 - 4 | 155 | 69 | |
| AK028955.1 | - | 3 - 4 | 161 | 69 | ||
| AK045539.1 | - | 3 - 4 | 233 | 69 | ||
| BC024599.1 | - | 3 - 4 | 203 | 5044495 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aif1l
Genomic Map