Assay Details
| Gene Symbol: Alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145362.2 | NP_663337.2 | 8 - 9 | 934 | 74 | |
| GenBank mRNA | AK145871.1 | - | 8 - 9 | 934 | 74 | |
| BC011281.1 | - | 7 - 8 | 841 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alg1
Genomic Map