Assay Details
| Gene Symbol: Rho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145383.1 | NP_663358.1 | 3 - 4 | 777 | 66 | |
| GenBank mRNA | AK044333.1 | - | 3 - 4 | 797 | 66 | |
| AK044412.1 | - | 3 - 4 | 795 | 66 | ||
| BC013125.1 | - | 3 - 4 | 777 | 4500760 | 66 | |
| BC031766.1 | - | 3 - 4 | 778 | 4527040 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rho
Genomic Map