Assay Details
| Gene Symbol: Prmt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145404.1 | NP_663379.1 | 3 - 4 | 256 | 66 | |
| GenBank mRNA | AK040103.1 | - | 3 - 4 | 288 | 66 | |
| AK154255.1 | - | 3 - 4 | 283 | 66 | ||
| AK173304.1 | - | 3 - 4 | 276 | 66 | ||
| AY673972.1 | - | 2 - 3 | 126 | 66 | ||
| BC006705.1 | - | 3 - 4 | 256 | 3583763 | 66 | |
| BC012845.1 | - | 2 - 3 | 77 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC012845.1 transcripts.
Set Membership: |
|

Prmt7
Genomic Map