Assay Details
| Gene Symbol: Prmt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145404.1 | NP_663379.1 | 14 - 15 | 1703 | 76 | |
| GenBank mRNA | AK154255.1 | - | 14 - 15 | 1730 | 76 | |
| AK162376.1 | - | 297 | 76 | |||
| AK173304.1 | - | 14 - 15 | 1723 | 76 | ||
| AY673972.1 | - | 13 - 14 | 1573 | 76 | ||
| BC006705.1 | - | 14 - 15 | 1703 | 3583763 | 76 | |
| BC057177.1 | - | 14 - 15 | 1727 | 5010547 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prmt7
Genomic Map