Assay Details
| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145409.2 | NP_663384.2 | 1 - 2 | 168 | 83 | |
| GenBank mRNA | AK077947.1 | - | 1 - 2 | 135 | 83 | |
| BB861615.1 | - | 1 - 2 | 178 | 84 | ||
| BC024142.1 | - | 1 - 2 | 155 | 5149957 | 83 | |
| BC083096.1 | - | 1 - 2 | 131 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chtf18
Genomic Map