Assay Details
| Gene Symbol: Chtf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145409.2 | NP_663384.2 | 4 - 5 | 675 | 62 | |
| GenBank mRNA | AK052673.1 | - | 4 - 5 | 634 | 62 | |
| AK077947.1 | - | 4 - 5 | 642 | 62 | ||
| AK181238.1 | - | 2 - 3 | 238 | 62 | ||
| BC024142.1 | - | 4 - 5 | 662 | 5149957 | 62 | |
| BC083096.1 | - | 4 - 5 | 638 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chtf18
Genomic Map