Assay Details
| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145416.3 | NP_663391.2 | 5 - 6 | 469 | 93 | |
| GenBank mRNA | BC021438.1 | - | 5 - 6 | 462 | 5039543 | 93 |
| CO426049.1 | - | 2 - 3 | 112 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kri1
Genomic Map