Assay Details
| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145416.3 | NP_663391.2 | 8 - 9 | 700 | 103 | |
| GenBank mRNA | BC021438.1 | - | 8 - 9 | 693 | 5039543 | 103 |
| CO426049.1 | - | 5 - 6 | 343 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kri1
Genomic Map