Assay Details
| Gene Symbol: Hkdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145419.1 | NP_663394.1 | 9 - 10 | 1362 | 62 | |
| GenBank mRNA | AK145980.1 | - | 9 - 10 | 1341 | 62 | |
| BC016235.1 | - | 9 - 10 | 1362 | 4504302 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hkdc1
Genomic Map