Assay Details
| Gene Symbol: Cd300ld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145437.2 | NP_663412.1 | 3 - 4 | 582 | 85 | |
| GenBank mRNA | AB292062.1 | - | 3 - 4 | 506 | 85 | |
| AK028662.1 | - | 3 - 4 | 594 | 85 | ||
| AK041292.1 | - | 3 - 4 | 618 | 85 | ||
| AY457051.1 | - | 3 - 4 | 506 | 85 | ||
| BC019814.1 | - | 3 - 4 | 582 | 5125383 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1983523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 442027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649879 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124223 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1983523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150768 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07501237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124223 | - | - |
| Whole Mouse Genome G4122A | A_51_P179789 | BC019814.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179789 | NM_145437.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179789 | AK028662.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131568 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111524 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09077821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09077814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08978751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09178974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08978754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09077821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150774 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10392809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392811 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424832_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10392808 | - | - |
| Mouse Genome 430 2.0 Array | 1424832_at | - | - |
| Mouse Genome 430 2.0 Array | 1424832_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217305_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217305_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100038028.mm.1 | NM_145437.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038028.mm.1 | NM_145437.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038028.mm.1 | TC1100004053.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038025.mm.1 | NM_145437.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038025.mm.1 | NM_145437.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038025.mm.1 | TC1100004053.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038026.mm.1 | NM_145437.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038026.mm.1 | NM_145437.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038026.mm.1 | TC1100004053.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038027.mm.1 | NM_145437.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038027.mm.1 | NM_145437.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038027.mm.1 | TC1100004053.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424832_at | - | - |
| Mouse Expression Set 430 | 1424832_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424832_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424832_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424832_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424832_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 427311 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7040497 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040497 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230287 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230287 | - | - |
| mouse-6 v1.1 (Illumina) | scl39351.4_153-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39351.4_153-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040497 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230287 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230287 | - | - |

Cd300ld
Genomic Map