Assay Details
Gene Symbol: Acot5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC016076.1 | - | 3 - 4 | 994 | 4485973 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28596200 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28596194 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038362 | AY563099.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038362 | BC016076.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038362 | NM_145444.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040260 | AY563099.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111256 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286615 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470338 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12951 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110174 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038362 | AY563099.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038362 | BC016076.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038362 | NM_145444.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040260 | AY563099.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P355996 | BC016076.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596194 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596202 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596208 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596210 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28596185 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110174 | - | - |
Whole Mouse Genome G4122A | A_51_P355996 | BC016076.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110771 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110174 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10105119 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10105116 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10105122 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28596194 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28596200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28596283 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10131258 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09833192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10294675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10216937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09759424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596165 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596202 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10217716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09817497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28596167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09759425 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10397168 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10397167 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10397169 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10397171 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10397170 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425257_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10397166 | - | - |
Mouse Genome 430 2.0 Array | 1425257_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1425257_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217698_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217698_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200005706.mm.1 | NM_145444.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005706.mm.1 | NM_145444.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200005706.mm.1 | TC1200000801.mm.1 | MTA TC |
Mouse Expression Set 430 | 1425257_at | - | - |
Mouse Expression Set 430 | 1425257_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425257_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425257_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425257_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425257_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 338492 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1230019 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001230019 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2718421 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2718421 | - | - |
mouse-6 v1.1 (Illumina) | scl42989.4.1_93-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1230019 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl42989.4.1_93-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230019 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2718421 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2718421 | - | - |
