| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400000698.mm.1 |
NM_145458.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000698.mm.1 |
NM_145458.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000698.mm.1 |
NM_178279.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000698.mm.1 |
NM_178279.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000698.mm.1 |
TC1400000076.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400000697.mm.1 |
NM_145458.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000697.mm.1 |
NM_145458.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000697.mm.1 |
NM_178279.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000697.mm.1 |
NM_178279.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400000697.mm.1 |
TC1400000076.mm.1 |
MTA TC |