Assay Details
| Gene Symbol: Haus4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145462.2 | NP_663437.2 | 2 - 3 | 261 | 84 | |
| GenBank mRNA | AK079161.1 | - | 2 - 3 | 201 | 84 | |
| AK079852.1 | - | 2 - 3 | 229 | 84 | ||
| AK087969.1 | - | 2 - 3 | 84 | |||
| BB855694.1 | - | 2 - 3 | 84 | |||
| BC004644.1 | - | 2 - 3 | 3157901 | 84 | ||
| BC011095.1 | - | 2 - 3 | 3157901 | 84 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK079161.1,AK079852.1,BC004644.1,BC011095.1,NM_145462.2 transcripts.
Set Membership: |
|

Haus4
Genomic Map