Assay Details
| Gene Symbol: Haus4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145462.2 | NP_663437.2 | 3 - 4 | 401 | 57 | |
| GenBank mRNA | AK079161.1 | - | 3 - 4 | 341 | 57 | |
| AK079852.1 | - | 3 - 4 | 369 | 57 | ||
| AK087969.1 | - | 3 - 4 | 376 | 57 | ||
| BC004644.1 | - | 3 - 4 | 376 | 3157901 | 57 | |
| BC011095.1 | - | 3 - 4 | 376 | 3157901 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Haus4
Genomic Map