Assay Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145476.2 | NP_663451.2 | 9 - 10 | 1238 | 89 | |
| GenBank mRNA | AK039627.1 | - | 9 - 10 | 1251 | 89 | |
| AK146872.1 | - | 9 - 10 | 1239 | 89 | ||
| AK154226.1 | - | 9 - 10 | 1260 | 89 | ||
| AK154602.1 | - | 9 - 10 | 1281 | 89 | ||
| AK155459.1 | - | 9 - 10 | 1252 | 89 | ||
| AK155770.1 | - | 9 - 10 | 1281 | 89 | ||
| BC023106.1 | - | 6 - 7 | 633 | 4016550 | 89 | |
| BC066009.1 | - | 9 - 10 | 1238 | 5708486 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d22a
Genomic Map