Assay Details
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145482.3 | NP_663457.2 | 9 - 10 | 1390 | 72 | |
GenBank mRNA | AK137485.1 | - | 1 - 2 | 1249 | 72 | |
AY037804.1 | - | 9 - 10 | 1390 | 72 | ||
BC029101.1 | - | 9 - 10 | 1905 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
