Assay Details
| Gene Symbol: Zfp160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145483.2 | NP_663458.2 | 4 - 5 | 399 | 107 | |
| GenBank mRNA | AK143313.1 | - | 4 - 5 | 400 | 107 | |
| AK173216.1 | - | 4 - 5 | 430 | 107 | ||
| BC015291.1 | - | 4 - 5 | 365 | 4242025 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31084705 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31084703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31084704 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049423 | AK173216.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049423 | BC015291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049423 | AK080975.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049423 | AK143313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2049423 | NM_145483.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339467 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234807 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 82591 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130103 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153384 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P949005 | BC040101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P949005 | AK161712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P949005 | AK162583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P949005 | BC040101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P949005 | AK161712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P949005 | AK162583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049423 | AK173216.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049423 | BC015291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049423 | AK080975.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049423 | AK143313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2049423 | NM_145483.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369796 | AK173216.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369796 | BC015291.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369796 | BE989157.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369796 | AK080975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369796 | NM_145483.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084695 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08733459 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03290547 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08733451 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130103 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153384 | - | - |
| Whole Mouse Genome G4122A | A_51_P369796 | AK173216.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369796 | BC015291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369796 | BE989157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369796 | AK080975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P369796 | NM_145483.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120912 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122593 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153384 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130103 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08733451 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13303259 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13327467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13303261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13303260 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13303254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13327465 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31084705 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31084704 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31084703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13303254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13416385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13416377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13102070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13527809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13416384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13303257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13102069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13416388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31084713 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442173 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452126_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442172 | - | - |
| Mouse Genome 430 2.0 Array | 1452126_at | - | - |
| Mouse Genome 430 2.0 Array | 1452126_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439073_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439073_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224585_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224585_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700001927.mm.1 | NM_145483.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700001927.mm.1 | NM_145483.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700001927.mm.1 | TC1700000257.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700001926.mm.1 | NM_145483.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700001926.mm.1 | NM_145483.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700001926.mm.1 | TC1700000257.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452126_PM_at | - | - |
| Mouse Expression Set 430 | 1452126_at | - | - |
| Mouse Expression Set 430 | 1439073_at | - | - |
| Mouse Expression Set 430 | 1439073_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452126_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452126_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452126_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452126_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 686887 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5570358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2513675 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2513675 | - | - |
| mouse-6 v1.1 (Illumina) | scl51065.1.493_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51065.1.493_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2867013 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2867013 | - | - |

Zfp160
Genomic Map