Assay Details
| Gene Symbol: Tkfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145496.1 | NP_663471.1 | 2 - 3 | 284 | 63 | |
| GenBank mRNA | AK138956.1 | - | 324 | 63 | ||
| AK171418.1 | - | 3 - 4 | 500 | 63 | ||
| BC021917.1 | - | 2 - 3 | 284 | 4481611 | 63 | |
More Information
Set Membership: |
|

Tkfc
Genomic Map