Assay Details
| Gene Symbol: Tkfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145496.1 | NP_663471.1 | 9 - 10 | 953 | 57 | |
| GenBank mRNA | AK138956.1 | - | 993 | 57 | ||
| AK171418.1 | - | 10 - 11 | 1144 | 57 | ||
| BC021917.1 | - | 9 - 10 | 953 | 4481611 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tkfc
Genomic Map