Assay Details
| Gene Symbol: Gca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145523.3 | NP_663498.1 | 1 - 2 | 131 | 74 | |
| GenBank mRNA | AF518325.1 | - | 1 - 2 | 115 | 74 | |
| AK033740.1 | - | 1 - 2 | 164 | 74 | ||
| AK046347.1 | - | 1 - 2 | 173 | 74 | ||
| AK049317.1 | - | 1 - 2 | 132 | 74 | ||
| AK050540.1 | - | 1 - 2 | 173 | 74 | ||
| AK078915.1 | - | 1 - 2 | 132 | 74 | ||
| BC021450.1 | - | 1 - 2 | 125 | 5044040 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF518325.1,AK033740.1,AK046347.1,AK049317.1,AK050540.1,AK078915.1,BC021450.1,NM_145523.3 transcripts.
Set Membership: |
|

Gca
Genomic Map