Assay Details
| Gene Symbol: Gca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145523.3 | NP_663498.1 | 4 - 5 | 417 | 94 | |
| GenBank mRNA | AB088388.1 | - | 4 - 5 | 311 | 94 | |
| AF518325.1 | - | 4 - 5 | 401 | 94 | ||
| AK033740.1 | - | 4 - 5 | 450 | 94 | ||
| AK046347.1 | - | 4 - 5 | 459 | 94 | ||
| AK049317.1 | - | 4 - 5 | 418 | 94 | ||
| AK050540.1 | - | 4 - 5 | 459 | 94 | ||
| AK078915.1 | - | 4 - 5 | 418 | 94 | ||
| BC021450.1 | - | 4 - 5 | 411 | 5044040 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gca
Genomic Map