Assay Details
| Gene Symbol: Gca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145523.3 | NP_663498.1 | 5 - 6 | 566 | 109 | |
| GenBank mRNA | AB088388.1 | - | 5 - 6 | 460 | 109 | |
| AF518325.1 | - | 5 - 6 | 550 | 109 | ||
| AK033740.1 | - | 5 - 6 | 599 | 109 | ||
| AK046347.1 | - | 5 - 6 | 608 | 109 | ||
| AK049317.1 | - | 5 - 6 | 567 | 109 | ||
| AK050540.1 | - | 5 - 6 | 608 | 109 | ||
| AK078915.1 | - | 5 - 6 | 567 | 109 | ||
| BC021450.1 | - | 5 - 6 | 560 | 5044040 | 109 | |
More Information
Set Membership: |
|

Gca
Genomic Map