Assay Details
| Gene Symbol: Atg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145528.3 | NP_663503.1 | 11 - 12 | 1282 | 91 | |
| GenBank mRNA | AK122339.1 | - | 7 - 8 | 3644 | 91 | |
| AK149672.1 | - | 11 - 12 | 1241 | 91 | ||
| AK154470.1 | - | 11 - 12 | 1270 | 91 | ||
| AK163962.1 | - | 11 - 12 | 1236 | 91 | ||
| AK171206.1 | - | 11 - 12 | 1268 | 91 | ||
| AK171440.1 | - | 11 - 12 | 1259 | 91 | ||
| BC016669.1 | - | 11 - 12 | 1266 | 2648797 | 91 | |
| BC023673.1 | - | 11 - 12 | 1232 | 5343236 | 91 | |
| BC023712.1 | - | 11 - 12 | 1255 | 5346303 | 91 | |
| BC033419.1 | - | 11 - 12 | 1233 | 5359944 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg13
Genomic Map