Assay Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293797.1 | NP_001280726.1 | 5 - 6 | 523 | 86 | |
| NM_145597.4 | NP_663572.1 | 6 - 7 | 627 | 86 | ||
| NR_122028.1 | 6 - 7 | 627 | 86 | |||
| GenBank mRNA | AK049979.1 | - | 6 - 7 | 626 | 86 | |
| AK082758.1 | - | 6 - 7 | 623 | 86 | ||
| AK082916.1 | - | 5 - 6 | 524 | 86 | ||
| AK088569.1 | - | 5 - 6 | 513 | 86 | ||
| AK147962.1 | - | 6 - 7 | 620 | 86 | ||
| BC021367.1 | - | 6 - 7 | 627 | 5030434 | 86 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P135130 | BC021367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P135130 | AK147962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P135130 | AK167517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P135130 | AK207333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P135130 | AK082758.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | AK147962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | AK049979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | AK032711.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | NM_145597.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | AK147962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | AK049979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | AK032711.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383218 | NM_145597.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79476 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111291 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115174 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | AK147962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | AK049979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | AK032711.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | NM_145597.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | AK147962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | AK049979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | AK032711.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383218 | NM_145597.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P135130 | BC021367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P135130 | AK147962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P135130 | AK167517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P135130 | AK207333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P135130 | AK082758.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25950232 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115174 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06547814 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111291 | - | - |
| Whole Mouse Genome G4122A | A_52_P590665 | BC021367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590665 | AK147962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590665 | AK167517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590665 | AK082758.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P590665 | AK082916.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | AK147962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | AK049979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | AK032711.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | NM_145597.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | AK147962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | AK049979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | AK032711.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383218 | NM_145597.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135130 | BC021367.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135130 | AK147962.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135130 | AK167517.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135130 | AK207333.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135130 | AK082758.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115174 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111291 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06703740 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06692316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25950248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07040602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06953151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06937289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06953156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07062692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06937297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06937298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06937305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06703743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07131291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07131289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07131292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07054580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25950260 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572325 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572326 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426211_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426212_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455705_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572318 | - | - |
| Mouse Genome 430 2.0 Array | 1426212_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455705_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426212_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455705_at | - | - |
| Mouse Genome 430 2.0 Array | 1426211_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426211_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234371_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234371_at | - | - |
| Mouse Expression Set 430 | 1455705_PM_at | - | - |
| Mouse Expression Set 430 | 1455705_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426211_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426212_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426211_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426212_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455705_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455705_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 743823 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4780563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223365 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223365 | - | - |
| mouse-6 v1.1 (Illumina) | scl000564.1_24-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000564.1_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2806740 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2806740 | - | - |

Tmem161a
Genomic Map