Assay Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293797.1 | NP_001280726.1 | 6 - 7 | 578 | 65 | |
| NM_145597.4 | NP_663572.1 | 7 - 8 | 682 | 65 | ||
| NR_122028.1 | 7 - 8 | 682 | 65 | |||
| GenBank mRNA | AK032711.1 | - | 3 - 4 | 198 | 65 | |
| AK049979.1 | - | 7 - 8 | 681 | 65 | ||
| AK082758.1 | - | 7 - 8 | 678 | 65 | ||
| AK082916.1 | - | 6 - 7 | 579 | 65 | ||
| AK088569.1 | - | 6 - 7 | 568 | 65 | ||
| AK147962.1 | - | 7 - 8 | 675 | 65 | ||
| AK167517.1 | - | 2 - 3 | 80 | 65 | ||
| BC021367.1 | - | 7 - 8 | 682 | 5030434 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem161a
Genomic Map