Assay Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293797.1 | NP_001280726.1 | 9 - 10 | 1017 | 58 | |
| NM_145597.4 | NP_663572.1 | 10 - 11 | 1121 | 58 | ||
| NR_122028.1 | 9 - 10 | 1026 | 58 | |||
| GenBank mRNA | AK032711.1 | - | 6 - 7 | 637 | 58 | |
| AK049979.1 | - | 10 - 11 | 1120 | 58 | ||
| AK082758.1 | - | 9 - 10 | 1022 | 58 | ||
| AK082916.1 | - | 9 - 10 | 1018 | 58 | ||
| AK088569.1 | - | 9 - 10 | 1007 | 58 | ||
| AK147962.1 | - | 10 - 11 | 1114 | 58 | ||
| AK167517.1 | - | 4 - 5 | 424 | 58 | ||
| BC021367.1 | - | 10 - 11 | 1121 | 5030434 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem161a
Genomic Map