Assay Details
| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145611.4 | NP_663586.3 | 3 - 4 | 1412 | 73 | |
| GenBank mRNA | AK031324.1 | - | 3 - 4 | 1405 | 73 | |
| AK045492.1 | - | 3 - 4 | 1384 | 73 | ||
| AK049288.1 | - | 3 - 4 | 1413 | 73 | ||
| BC006647.1 | - | 3 - 4 | 1400 | 73 | ||
| BC010245.1 | - | 3 - 4 | 1369 | 3710765 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kank2
Genomic Map