Assay Details
| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145611.4 | NP_663586.3 | 9 - 10 | 2334 | 103 | |
| GenBank mRNA | AK031324.1 | - | 9 - 10 | 2327 | 103 | |
| AK045492.1 | - | 9 - 10 | 2306 | 103 | ||
| AK049288.1 | - | 9 - 10 | 2335 | 103 | ||
| BC006647.1 | - | 9 - 10 | 2322 | 103 | ||
| BC010245.1 | - | 8 - 9 | 2203 | 3710765 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kank2
Genomic Map