Assay Details
| Gene Symbol: Zfp810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145612.2 | NP_663587.1 | 5 - 6 | 507 | 91 | |
| GenBank mRNA | AK032556.1 | - | 5 - 6 | 547 | 91 | |
| BC005471.1 | - | 5 - 6 | 507 | 3496918 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp810
Genomic Map