| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900020450.mm.1 |
NM_001146046.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020450.mm.1 |
NM_001146046.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020450.mm.1 |
NM_145616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020450.mm.1 |
NM_145616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020450.mm.1 |
TC0900002435.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020452.mm.1 |
NM_001146046.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020452.mm.1 |
NM_001146046.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020452.mm.1 |
NM_145616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020452.mm.1 |
NM_145616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020452.mm.1 |
TC0900002435.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900020447.mm.1 |
NM_001146046.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020447.mm.1 |
NM_001146046.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020447.mm.1 |
NM_001146047.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020447.mm.1 |
NM_001146047.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020447.mm.1 |
NM_145616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020447.mm.1 |
NM_145616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900020447.mm.1 |
TC0900002435.mm.1 |
MTA TC |