Assay Details
| Gene Symbol: Lrrc49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146046.1 | NP_001139518.1 | 4 - 5 | 572 | 90 | |
| NM_001146047.1 | NP_001139519.1 | 5 - 6 | 571 | 90 | ||
| NM_145616.4 | NP_663591.3 | 5 - 6 | 770 | 90 | ||
| GenBank mRNA | AK041627.1 | - | 4 - 5 | 341 | 90 | |
| AK049268.1 | - | 5 - 6 | 549 | 90 | ||
| AK051325.1 | - | 4 - 5 | 377 | 90 | ||
| AK085022.1 | - | 565 | 91 | |||
| BC016574.1 | - | 4 - 5 | 339 | 3594577 | 90 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK085022.1 transcripts.
Set Membership: |
|

Lrrc49
Genomic Map