Assay Details
| Gene Symbol: Camkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145621.2 | NP_663596.1 | 4 - 5 | 446 | 69 | |
| GenBank mRNA | AK085143.1 | - | 452 | 69 | ||
| AK147323.1 | - | 4 - 5 | 434 | 69 | ||
| BC017634.1 | - | 4 - 5 | 446 | 4527857 | 69 | |
| BC111033.1 | - | 4 - 5 | 441 | 5368225 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camkv
Genomic Map