Assay Details
| Gene Symbol: Camkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145621.2 | NP_663596.1 | 7 - 8 | 781 | 72 | |
| GenBank mRNA | AK085143.1 | - | 786 | 72 | ||
| AK147323.1 | - | 7 - 8 | 769 | 72 | ||
| BC017634.1 | - | 7 - 8 | 781 | 4527857 | 72 | |
| BC022169.1 | - | 5 - 6 | 1147 | 72 | ||
| BC086762.1 | - | 6 - 7 | 979 | 72 | ||
| BC111033.1 | - | 7 - 8 | 776 | 5368225 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camkv
Genomic Map