Assay Details
| Gene Symbol: Alox12e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145684.1 | NP_663717.1 | 3 - 4 | 478 | 87 | |
| GenBank mRNA | AK138743.1 | - | 3 - 4 | 432 | 87 | |
| BC013751.1 | - | 3 - 4 | 458 | 3596809 | 87 | |
| BC051047.1 | - | 3 - 4 | 462 | 4457881 | 87 | |
| U39200.1 | - | 3 - 4 | 419 | 87 | ||
| X99252.1 | - | 3 - 4 | 478 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox12e
Genomic Map