Assay Details
| Gene Symbol: Ackr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145700.2 | NP_663746.2 | 1 - 2 | 222 | 86 | |
| GenBank mRNA | AY072938.1 | - | 1 - 2 | 514 | 86 | |
| BC140999.1 | - | 1 - 2 | 222 | 9055611 | 86 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY072938.1,BC140999.1,NM_145700.2 transcripts.
Set Membership: |
|

Ackr4
Genomic Map