Assay Details
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146148.2 | NP_666260.1 | 1 - 2 | 119 | 113 | |
GenBank mRNA | AB077295.1 | - | 1 - 2 | 133 | 113 | |
AB077296.1 | - | 1 - 2 | 82 | 113 | ||
AB077298.1 | - | 1 - 2 | 82 | 113 | ||
AK090245.1 | - | 1 - 2 | 117 | 113 | ||
BC027748.1 | - | 1 - 2 | 115 | 5052412 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
