Assay Details
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290645.1 | NP_001277574.1 | 6 - 7 | 1042 | 104 | |
NM_146148.2 | NP_666260.1 | 7 - 8 | 1139 | 104 | ||
GenBank mRNA | AB077295.1 | - | 7 - 8 | 1153 | 104 | |
AB077296.1 | - | 7 - 8 | 1102 | 104 | ||
AB077297.1 | - | 6 - 7 | 1008 | 104 | ||
AB077298.1 | - | 7 - 8 | 1102 | 104 | ||
AK090245.1 | - | 7 - 8 | 1139 | 104 | ||
BC027748.1 | - | 7 - 8 | 1135 | 5052412 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
